Organic reference standard, alpha-Terpineol, analytical standard, Supelco®
Lieferant: Merck
30627-100MG
30627-100MGEA 58.4 EUR
30627-100MG
Organic reference standard, alpha-Terpineol, analytical standard, Supelco®
Standards Organische Standards
α-Terpineol is a monoterpene alcohol. It is one of the components responsible for the antifungal activity of Melaleuca alternifolia (tea tree) essential oil.
The reaction rate constant of α-terpineol with the OH radical and ozone was found to be (1.9±0.5)×10⁻¹⁰ cm³ molecule⁻¹s⁻¹ and (3.0±0.2)×10⁻¹⁶ cm³ molecule⁻¹s⁻¹, respectively.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b10207ec3e5d5c07b613dae537e0e9028887067&vid=0b10207ec3e5d5c07b613dae537e0e9028887067&prid=web&clid=SAPDAM