Reactive black 5 ≥50 % (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

306452-100G
306452-100GEA 132 EUR
306452-100G
Reactive black 5 ≥50 % (Farbstoffgehalt), Sigma-Aldrich®
Reactive black 5

Reactive Black 5 (RB5) has been: used in an optimization study on the photocatalytic degradation of RB5 dye in synthetic greywater effluent (i.e. aqueous RB5 in distilled water), in order to understand the effects of different photoreactor parameters on the degradation kinetics. As a textile dye to investigate the anaerobic reduction in upflow stirred packed-bed reactors (USPBRs) with biological activated carbon (BAC) system. Used in the performance evaluation of dye removal forlarge molecule substance.

Formel: C₂₆H₂₁N₅Na₄O₁₉S₆
Schmelzpunkt: >300 °C (Lit.)
Dichte: 1,21 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
CAS-Nummer: 17095-24-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=410db0c85e1e68760544d2f526d991fa26b0cfa5&vid=48763a15cab555c402cf64b99f18cf2921a65b8d&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR