Blei(II)nitrat 0.1 mol/l (0.2 N) aqueous solution
Lieferant: VWR Chemicals
|
|
Synonyme:
Bleidinitrat
307227.0500EA
41.1
EUR
307227.0500
Blei(II)nitrat 0.1 mol/l (0.2 N) aqueous solution
Blei(II)nitrat
Formel:
Pb(NO₃)₂ Molecular Weight: 331.21 g/mol |
MDL:
MFCD00011153 CAS-Nummer: 10099-74-8 EINECS: 233-245-9 UN: 3082 ADR: 9 III Merck Index: 13,05432 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...