Anthron ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
CAS No.: 90-44-8
Molecular Formula: C14H10O
Formula Weight: 194.23
Physical Form: Crystalline
Sensitivity: Light Sensitive
Melting Point: 155-158°
Beilstein Reference: 1910173
Merck Reference: 14,691
MDL No.: MFCD00001187
Used for determination of carbohydrates
Formel:
C₁₄H₁₀O Schmelzpunkt: 155…158 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001187 CAS-Nummer: 90-44-8 EINECS: 201-994-0 Merck Index: 14,691 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...