1,2-Dimethoxyethan ≥99,9% inhibitorfrei, CHROMASOLV™ für die HPLC, Riedel-de Haën™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Ethylenglycoldimethylether, EGDME, Dimethylglycol, DME
307432-1L
307432-1LEA
274.36
EUR
307432-1L
1,2-Dimethoxyethan ≥99,9% inhibitorfrei, CHROMASOLV™ für die HPLC, Riedel-de Haën™
1,2-Dimethoxyethan
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Formel:
C₄H₁₀O₂ Molecular Weight: 90,12 g/mol Siedepunkt: 85 °C (1,013 hPa) Schmelzpunkt: –58 °C Dichte: 0,87 g/cm³ (20 °C) Flammpunkt: –2 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008502 CAS-Nummer: 110-71-4 EINECS: 203-794-9 UN: 2252 ADR: 3,II Merck Index: 13,03251 |
Spezifikation Testergebnisse
Identity (IR) | complying |
Assay (GC) | Min. 99.85 % |
Non-volatile matter | Max. 0.0005 % |
Water (Karl Fischer) | Max. 0.03 % |
Peroxides (as H₂O₂) | Max. 15 ppm |
Absorbance at 400 nm | Max. 0.01 |
Absorbance at 350 nm | Max. 0.01 |
Absorbance at 300 nm | Max. 0.03 |
Absorbance at 250 nm | Max. 0.20 |
Absorbance at 220 nm | Max. 1.0 |
Vapor Density | 3.1 (20 °C, vs air) |
Refractive Index | n20/D 1.379 (lit.) |
Explosion Limit | 10.4 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...