Propylenoxid ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (±)-Propylenoxid, (±)-Propenoxid, (±)-Methyloxiran

30765.AE 30765.K2 30765.L7
30765.AEEA 27.6 EUR
30765.AE 30765.K2 30765.L7
Propylenoxid ≥99%
Propylenoxid

Purity: 99+% CAS No.: 75-56-9 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -37°(-35°F) Physical Form: Liquid Density: 0.83 Refractive Index: 1.3660 Appearance: Colourless Odour: Ether-like Melting Point: -112° Boiling Point: 34-35° Beilstein Reference: 79763 Merck Reference: 14,7856 UN No.: 1280 MDL No.: MFCD00005126

Formel: C₃H₆O
Siedepunkt: 34…35 °C
Schmelzpunkt: –112 °C
Dichte: 0,83
Flammpunkt: –37 °C (–35 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00005126
CAS-Nummer: 75-56-9
EINECS: 200-879-2
UN: 1280
ADR: 3,I
Merck Index: 14,7856
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=443494ff0620e18a4c5de7f3f4cd87d6e5eec44c&vid=33bc95a664261fbc904256d1b87a1972dde5dcd1&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR