Propylenoxid ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
(±)-Propylenoxid, (±)-Propenoxid, (±)-Methyloxiran
30765.AE
30765.K2
30765.L7
30765.AEEA
27.6
EUR
30765.AE
30765.K2
30765.L7
Propylenoxid ≥99%
Propylenoxid
Purity: 99+%
CAS No.: 75-56-9
Molecular Formula: C3H6O
Formula Weight: 58.08
Flash Point: -37°(-35°F)
Physical Form: Liquid
Density: 0.83
Refractive Index: 1.3660
Appearance: Colourless
Odour: Ether-like
Melting Point: -112°
Boiling Point: 34-35°
Beilstein Reference: 79763
Merck Reference: 14,7856
UN No.: 1280
MDL No.: MFCD00005126
Formel:
C₃H₆O Siedepunkt: 34…35 °C Schmelzpunkt: –112 °C Dichte: 0,83 Flammpunkt: –37 °C (–35 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00005126 CAS-Nummer: 75-56-9 EINECS: 200-879-2 UN: 1280 ADR: 3,I Merck Index: 14,7856 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...