Tetramethylammoniumhydroxid 25% w/w methanol
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
30833.09
30833.22
30833.36
30833.09EA
33.9
EUR
30833.09
30833.22
30833.36
Tetramethylammoniumhydroxid 25% w/w methanol
Tetramethylammonium hydroxide
CAS No.: 75-59-2
Molecular Formula: C4H13NO
Formula Weight: 91.15
Flash Point: 12°(54°F)
Physical Form: Liquid
Density: 0.865
Appearance: Colourless to pale yellow
Odour: Like alcohol
Merck Reference: 14,9224
UN No.: 2924
MDL No.: MFCD00008280
Formel:
C₄H₁₃NO Molecular Weight: 91.15 g/mol Dichte: 0.865 Flammpunkt: 12 °C (54 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008280 CAS-Nummer: 75-59-2 EINECS: 200-882-9 UN: 2924 ADR: 3 II Merck Index: 14,9224 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...