1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecan

Lieferant: Thermo Scientific
Achtung

Synonyme: 1,4,8,11-Tetramethylcyclam

30854.03
30854.03EA 224 EUR
30854.03
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecan
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane

CAS No.: 41203-22-9 Molecular Formula: C14H32N4 Formula Weight: 256.43 Flash Point: 110°(230°F) Physical Form: Crystalline Sensitivity: Hygroscopic Melting Point: 34-36° MDL No.: MFCD00005106

Formel: C₁₄H₃₂N₄
Molecular Weight: 256,44 g/mol
Schmelzpunkt: 34…36 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005106
CAS-Nummer: 41203-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR