Thioharnstoff ≥99,999% (Metall-Basis), Supelco®
Lieferant: Merck
Synonyme:
Aminomercaptoameisensäureamid, Thiocarbamid
30885-5G
30885-5GEA
219
EUR
30885-5G
Thioharnstoff ≥99,999% (Metall-Basis), Supelco®
Thioharnstoff
Thiourea is a chaotropic agent; strong denaturant. Increases solubility and recovery of proteins.
Formel:
NH₂CSNH₂ Molecular Weight: 76.12 g/mol |
MDL:
MFCD00008067 CAS-Nummer: 62-56-6 Merck Index: 13,09443 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...