Quinoline yellow sulphonic acid sodium salt (Acid yellow 3), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

309052-25G
309052-25GEA 177 EUR
309052-25G
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3), Sigma-Aldrich®
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3)

Quinoline Yellow, Mixture of the mono- and disulfonic acids of Quinoline Yellow (Acid Yellow 3)

Formel: C₁₈H₁₁NO₂.xNaO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00080727
CAS-Nummer: 8004-92-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8874ee7e4dcbfe90964cb38be2749316062b2237&vid=72fb321308f2469d26dfc52ef14f63865d84a154&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR