Organic reference standard, Rebaudioside C, analytical standard, Supelco®
Lieferant: Merck
30987-10MG
30987-10MGEA 710 EUR
30987-10MG
Organic reference standard, Rebaudioside C, analytical standard, Supelco®
Standards Organische Standards
Rebaudioside C is classified under the steviol glycosides group of compounds.
Rebaudioside C may be used as a reference standard for the determination of rebaudioside C in, Soft and alcoholic beverages by high-performance liquid chromatography and tandem mass spectrometry with electrospray ionization (HPLC-ESI-MS/MS). Leaves of Stevia rebaudiana (Bertoni) from plants cultivated under chilling stress conditions by untargeted LC-ESI (Orbitrap) MS, LC-ESI coupled with triple quadrupole (QqQ) MS/MS and multivariate data analysis. Stevia formulations and sugar-free food products by high performance thin layer chromatography (HPTLC) with ESI-MS, hydrophilic interaction chromatography (HILIC)-HPLC as well as reversed phase (RP)-HPLC. Popular beverages by HILIC-MS/MS.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=982e55884301742657301a71744b887591a6aaa8&vid=982e55884301742657301a71744b887591a6aaa8&prid=web&clid=SAPDAM