Quecksilber(II)chlorid ≥99,5% (Trockenmasse), Puriss. p.a. ACS, ISO, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

31005-500G
31005-500GEA 513.66 EUR
31005-500G
Quecksilber(II)chlorid ≥99,5% (Trockenmasse), Puriss. p.a. ACS, ISO, Fluka™
Quecksilber(II)chlorid
Formel: HgCl₂
Siedepunkt: 302 °C
Schmelzpunkt: 276 °C
Dichte: 5,44 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011041
CAS-Nummer: 7487-94-7
EINECS: 231-299-8
UN: 1624
ADR: 6.1,II
Merck Index: 13,05901

Bestellen

Spezifikation Testergebnisse

Assay (calculated to the dried substance) Min. 99.5 %
Loss on drying (on P₂O₅) Max. 1.0 %
Residue on ignition a. reducing with HCOOH (SO₄)* Max. 0.02 %
Water insoluble matter Max. 0.01 %
Silver (Ag) Max. 0.0005 %
Calcium (Ca) Max. 0.001 %
Cadmium (Cd) Max. 0.0005 %
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 0.001 %
Iron (Fe) Max. 0.0005 %
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.001 %
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 0.005 %
Nickel (Ni) Max. 0.0005 %
Lead (Pb) Max. 0.001 %
Zinc (Zn) Max. 0.0005 %
Other heavy metals (as Pb) Max. 0.001 %
Mercury(I) chloride Max. 0.05 %
Total S (as SO₄) Max. 50 ppm
Sulfate (SO₄) Max. 0.02 %
Total N Max. 0.002 %
In ether insoluble matter complying
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR