Diethyl-3,4-pyridinedicarboxylate ≥97%, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
Synonyme:
Cinchomeronic ester, diethyl pyridine-3,4-dicarboxylate, ethyl 4-(ethoxycarbonyl)pyridine-3-carboxylate
310212-5G
310212-5GEA
210
EUR
310212-5G
Diethyl-3,4-pyridinedicarboxylate ≥97%, Sigma-Aldrich®
Diethyl-3,4-pyridinedicarboxylate
Diethyl 3,4-pyridinedicarboxylate,97% (Cinchomeronic ester)
Formel:
C₁₁H₁₃NO₄ Molecular Weight: 223,228 g/mol Dichte: 1,145 g/mL (25 °C) (Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012349 CAS-Nummer: 1678-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...