Yeast vitality staining kit
Lieferant: Biotium
31062
31062.EA 385 EUR
31062.
Yeast vitality staining kit
Tests Zelltests
A fluorescent vitality dye for yeast, identical in structure to the dye FUN® 1. Living yeast cells show red fluorescent vacuolar tubules.
  • For live-cell imaging of yeast and fungi
  • ViaVac™ Red/Green stains the vacuole red in metabolically active cells
  • Calcofluor White labels all cell walls blue
The Yeast Vitality Staining Kit is a fluorogenic assay that allows you to determine whether your yeast cells are metabolically active. The kit contains two fluorescent components: Calcofluor White (BTIU29067) and ViaVac™ Red/Green (BTIU29068).
ViaVac™ Red/Green (BTIU29068) is a fluorescent viability dye for yeast, identical in structure to the dye FUN® 1. ViaVac™ passively diffuses into cells, showing green cytoplasmic staining. In many species of yeast and fungi, in metabolically active cells the dye is actively transported into the vacuole, resulting in red fluorescent staining of tubular intravacuolar structures.
Calcofluor White (BTIU29067) is a fluorescent blue dye that binds to cellulose and chitin, which can be found in the cell walls of fungi, algae, and plants.
After staining yeast with this kit, both living and dead cells will be outlined in blue, while only living cells will show red fluorescent, vacuolar tubules.
Spectral Properties
ViacVac™ Red/Green Ex/Em ~485/620 nm (red); 485/530 nm (green)
Calcofluor White Ex/Em 360/430 nm
FUN®1 is a registered trademark of Thermo Fisher Scientific.
Lieferumfang: Kit contains ViaVac™ Red/Green, 10 mM in DMSO, 100 µl and Calcofluor White, 5 mM in water, 500 µl.