dsDNA quantitation kits for the Qubit® fluorometer, AccuGreen™, high-sensitivity and broad-range
Lieferant: Biotium
AccuGreen™ kit is specific for dsDNA and can quantify DNA samples in the range of 0,1 to 10 ng/μl.
- Optimised for use with the handheld fluorometers such as the Qubit® fluorometer
- Uses the pre-programmed Qubit® dsDNA high sensitivity assay setting
- Linear range in assay: 0,01 to 100 ng dsDNA
- Green fluorescence (Ex/Em 502/523 nm)
- Storage at 4 °C
Lieferumfang: Kit contains AccuGreen™ dye 200X, AccuGreen™ buffer 1X, AccuGreen™ standard 1 (0 ng/l) and AccuGreen™ standard 2 (10 ng/l).