Potassium trioxalatoferrate(III) trihydrate
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ferrate(3-), tris[ethanedioato(2-)-κO1,κO2]-, potassium, hydrate (1:3:3), (OC-6-11)-, Potassium iron(III) oxalate
31124.14
31124.22
31124.06
31124.36
31124.14EA
89.3
EUR
31124.14
31124.22
31124.06
31124.36
Potassium trioxalatoferrate(III) trihydrate
Potassium trioxalatoferrate(III) trihydrate
CAS No.: 5936-11-8
Molecular Formula: K3Fe(C2O4)3.3H2O
Formula Weight: 491.26 (437.21anhy)
Physical Form: Crystalline
Density: 2.133
Sensitivity: Hygroscopic
Appearance: Green
UN No.: UN3288
MDL No.: MFCD00150450
Formel:
K₃Fe(C₂O₄)₃·3H₂O Dichte: 2,133 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150450 CAS-Nummer: 5936-11-8 EINECS: 238-954-7 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...