Essigsäure (Eisessig) 99,5-100,5% USP, FCC, ACS, Macron Fine Chemicals™

Lieferant: Avantor

Synonyme: Ethansäure

C3121-04EA 63.45 EUR
C3121-04 3121-46
Essigsäure (Eisessig) 99,5-100,5% USP, FCC, ACS, Macron Fine Chemicals™
Essigsäure

Bitte beachten Preserve in tight containers, and store at room temperature

Formel: H₃CCOOH
Molecular Weight: 60,05 g/mol
Siedepunkt: 118 °C (1013 hPa)
Schmelzpunkt: 17 °C
Dichte: 1,05 g/cm³ (20 °C)
Flammpunkt: 39,4 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00036152
CAS-Nummer: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets U.S.P Requirements
Meets ACS Reagent Chemical Requirements
Meets F.C.C. Requirements
GMP Manufactured Product
Food GMP Manufactured Product
IMPORTANT: Material will freeze if stored below 17 °C (63°F).
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (CH₃COOH) 99.5 - 100.5 %
USP - Chloride (Cl) (USP) Passes Test
USP - Congealing Temperature ≥15.6 °C
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Readily Oxidizable Substances Passes Test
USP - Sulfate (USP)(SO₄) Passes Test
USP - Limit of Nonvolatile Residue ≤0.005 %
FCC - Assay (CH₃COOH) 99.7 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤0.5 mg/kg
FCC - Limit of Nonvolatile Residue ≤0.005 %
FCC - Readily Oxidizable Substances Passes Test
FCC - Solidification Point ≥15.6 °C
ACS - Assay (by GC, corrected for water) ≥99.7 %
ACS - Acetic Anhydride ((CH₃CO)₂O) ≤0.01 %
ACS - Chloride (Cl) ≤1 ppm
ACS - Color (APHA) ≤10
Dilution Test Passes Test
Trace Impurities - ACS - Iron (Fe) ≤0.200 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤0.5 ppm
ACS - Substances Reducing Dichromate Passes Test
ACS - Substances Reducing Permanganate Passes Test
ACS - Sulfate (SO₄) ≤1 ppm
ACS - Titrable Base (meq/g) ≤0.0004
Copper (Cu) ≤0.00001 %
Nickel (Ni) ≤0.00001 %
Residue after Evaporation ≤0.001 %
Appearance (clear, colorless liquid)
Residual Solvents: No Class 1, 2 or 3 residual solvents are used in or
produced by the production process.
Preserve in tight containers, and store at room temperature
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR