Kaliumnitrat ≥99%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO analytical reagent, Fluka™
Lieferant: Honeywell Chemicals
|
Achtung
|
Synonyme:
Kalisalpeter
31263-50KG
31263.9050
31263-100G
31263-1KG
31263-4X5KG
31263-500G
31263-6X1KG
31263-50KGEA
1810.59
EUR
31263-50KG
HONE31263.9050
31263-100G
31263-1KG
31263-4X5KG
31263-500G
31263-6X1KG
Kaliumnitrat ≥99%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO analytical reagent, Fluka™
Kaliumnitrat
Formel:
KNO₃ Siedepunkt: Decomposition Schmelzpunkt: 335 °C (1,013 hPa) Dichte: 2,11 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011409 CAS-Nummer: 7757-79-1 EINECS: 231-818-8 UN: 1486 ADR: 5.1,III REACH: 01-2119488224-35 Merck Index: 13,07733 |
Spezifikation Testergebnisse
Assay | Min. 99.0 % |
pH (5 %, 25 °C) | 5.0 - 8.0 |
Water insoluble matter | Max. 0.005 % |
Ammonium (NH₄) | Max. 0.001 % |
Calcium (Ca) | Max. 0.001 % |
Copper (Cu) | Max. 0.0005 % |
Iron (Fe) | Max. 2 ppm |
Magnesium (Mg) | Max. 0.002 % |
Sodium (Na) | Max. 0.005 % |
Lead (Pb) | Max. 0.0005 % |
Zinc (Zn) | Max. 0.0005 % |
Heavy metals (as Pb) | Max. 5 ppm |
Chloride (Cl) | Max. 0.001 % |
Iodate (IO₃) | Max. 5 ppm |
Nitrite (NO₂) | Max. 0.001 % |
Phosphate (PO₄) | Max. 5 ppm |
Sulfate (SO₄) | Max. 0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...