Milchsäure 88%, AR®, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

3130-46EA 426.26 EUR
3130-46
Milchsäure 88%, AR®, Macron Fine Chemicals™
Milchsäure
Formel: H₃CCH(OH)COOH
MDL: MFCD00004520
CAS-Nummer: 50-21-5
UN: 3265
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance colorless to light yellow, syrupy liquid Passes test
Assay 88 - 92 %
Reducing sugars passes test passes test Passes test
Residue after Ignition (as SO4) ≤ 0,04 %
Readily Carbonizable Substances Passes test
Chloride (Cl) ≤ 0,002 %
Sulfate (SO4) ≤ 0,005 %
Heavy Metals (as Pb) ≤ 0,0006 %
Arsenic (As) ≤ 0,0001 %
Iron (Fe) ≤ 0,0005 %
Stereochemical purity (L isomer assay) ≥95%
For Laboratory, Research or Manufacturing Use.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR