n-Dodecan ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99%
CAS No.: 112-40-3
Molecular Formula: C12H26
Formula Weight: 170.34
Flash Point: 71°(159°F)
Physical Form: Liquid
Density: 0.749
Refractive Index: 1.4220
Appearance: Colourless
Odour: Petrol-like
Melting Point: -10°
Boiling Point: 215-217°
Beilstein Reference: 1697175
MDL No.: MFCD00008969
Formel:
H₃C(CH₂)₁₀CH₃ Siedepunkt: 215…217 °C Schmelzpunkt: –10 °C Dichte: 0,749 Flammpunkt: 71 °C (159 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008969 CAS-Nummer: 112-40-3 EINECS: 203-967-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...