n-Dodecan ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

31324.14 31324.22 31324.36
31324.14EA 26.9 EUR
31324.14 31324.22 31324.36
n-Dodecan ≥99%
n-Dodecan

Purity: 99% CAS No.: 112-40-3 Molecular Formula: C12H26 Formula Weight: 170.34 Flash Point: 71°(159°F) Physical Form: Liquid Density: 0.749 Refractive Index: 1.4220 Appearance: Colourless Odour: Petrol-like Melting Point: -10° Boiling Point: 215-217° Beilstein Reference: 1697175 MDL No.: MFCD00008969

Formel: H₃C(CH₂)₁₀CH₃
Siedepunkt: 215…217 °C
Schmelzpunkt: –10 °C
Dichte: 0,749
Flammpunkt: 71 °C (159 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008969
CAS-Nummer: 112-40-3
EINECS: 203-967-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=616d3d75a63f52fbe2a8be1681d9233be6dc0928&vid=3268fe23453ad703df2782bd3c7ba6b939ba00cc&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR