Schwefelsäure ≥95%, AR®, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
|
Gefahr
|
3133-46EA
354.16
EUR
3133-46
Schwefelsäure ≥95%, AR®, Macron Fine Chemicals™
Schwefelsäure
Formel:
H₂SO₄ Molecular Weight: 98,08 g/mol |
MDL:
MFCD00064589 CAS-Nummer: 7664-93-9 |
Spezifikation Testergebnisse
Appearance | Clear, Colourless, Oily Liquid |
Assay | ≥95% |
Residue after Ignition | ≤0.001% |
Substances Reducing KMnO₄ (as SO₂) | ≤0.0003% |
Nitrate (NO₃) | ≤0.0002% |
Chloride (Cl) | ≤0.0001% |
Ammonium (NH₄) | ≤0.0003% |
Heavy Metals (as Pb) | ≤0.0002% |
For Laboratory, Research or Manufacturing Use | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...