Kaliumhydroxid ≥85,0%, Plätzchen, BAKER ANALYZED®, elektronische Qualität for Microelectronic, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Synonyme:
Kalihydrat, Ätzkali
3141-05EA
301
EUR
3141-05
3141-01
3141-07
3141-19
Kaliumhydroxid ≥85,0%, Plätzchen, BAKER ANALYZED®, elektronische Qualität for Microelectronic, J.T.Baker®
Kaliumhydroxid
Formel:
KOH Molecular Weight: 56.11 g/mol Siedepunkt: 1320 °C (1013 hPa) Schmelzpunkt: 360 °C Dichte: 2.04 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003553 CAS-Nummer: 1310-58-3 EINECS: 215-181-3 UN: 1813 ADR: 8 II REACH: 01-2119487136-33 Merck Index: 12,07806 |
Spezifikation Testergebnisse
For Microelectronic Use | ≥85.0% |
Assay (KOH) | ≥85.0 % |
Potassium Carbonate (K₂CO₃) | ≤1.0 % |
Insoluble Matter | ≤50 ppm |
Chloride (Cl) | ≤0.01 % |
Sulfate (SO₄) | ≤20 ppm |
Ammonium Hydroxide Precipitate | ≤0.02 % |
Sodium (Na) | ≤0.05 % |
Nitrogen Compounds (as N) | ≤10 ppm |
Phosphate (PO₄) | ≤5 ppm |
Heavy Metals (as Ag) | ≤10 ppm |
Iron (Fe) | ≤10 ppm |
Mercury (Hg), For Information Only | ≤10 ppm |
Nickel (Ni) | ≤10 ppm |
Calcium (Ca) | ≤50 ppm |
Magnesium (Mg), For Information Only |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...