Natriumcarbonat, wasserfrei ≥99,8% (Trockenmasse), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Fluka™

Lieferant: Honeywell Chemicals
Achtung

Synonyme: wasserfreie Soda

31432.9025 31432-1KG 31432-2.5KG 31432-500G 31432-5KG 31432-6X1KG 31432-250G 31432-25KG
HONE31432.9025EA 0 EUR
HONE31432.9025 31432-1KG 31432-2.5KG 31432-500G 31432-5KG 31432-6X1KG 31432-250G 31432-25KG
Natriumcarbonat, wasserfrei ≥99,8% (Trockenmasse), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Fluka™
Natriumcarbonat
Formel: Na₂CO₃
Molecular Weight: 105.99 g/mol
Siedepunkt: 1600
Schmelzpunkt: 854 °C
Dichte: 2.53 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003494
CAS-Nummer: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (calculated to the dried substance) Min. 99.8 %
Loss on drying (300°C) Max. 0.5 %
Water insoluble matter Max. 0.01 %
Silver (Ag) Max. 0.0005 %
Aluminium (Al) Max. 0.001 %
Arsenic (As) Max. 0.00005 %
Barium (Ba) Max. 0.0005 %
Bismuth (Bi) Max. 0.0005 %
Calcium (Ca) Max. 0.005 %
Cadmium (Cd) Max. 0.0005 %
Cobalt (Co) Max. 0.0005 %
Chromium (Cr) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 5 ppm
Lithium (Li) Max. 0.0005 %
Potassium (K) Max. 0.005 %
Manganese (Mn) Max. 0.0005 %
Magnesium (Mg) Max. 0.002 %
Molybdenum (Mo) Max. 0.0005 %
Nickel (Ni) Max. 0.0005 %
Lead (Pb) Max. 0.0005 %
Strontium (Sr) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 0.0005 %
Heavy metals (ICP) ACS Max. 5 ppm
Chloride (Cl) Max. 0.001 %
Total N Max. 0.001 %
Total S (as SO₄) Max. 0.003 %
Phosphate (PO₄) Max. 0.001 %
Silicate (as SiO₂) Max. 0.002 %
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR