Kaliumpermanganat 0,02 mol/l 0,1 N aqueous solution, AVS TITRINORM Volumetric standardised against ferrous ethylenediammonium sulphate

Lieferant: VWR Chemicals

Synonyme: Permanganic acid potassium salt

31456.295EA 56.6 EUR
31456.295
Kaliumpermanganat 0,02 mol/l 0,1 N aqueous solution, AVS TITRINORM Volumetric standardised against ferrous ethylenediammonium sulphate
Potassium permanganate
Formel: KMnO₄
Molecular Weight: 158.03 g/mol
MDL: MFCD00011364
CAS-Nummer: 7722-64-7
UN: 3082
ADR: 9 III
Merck Index: 12,07824

Bestellen

Spezifikation Testergebnisse

Titer (20°C; real value 0.2% accuracy) 0.01996 - 0.02004 mol/L
NIST traceable Confirmed
Conforms to Reag.Ph.Eur. Passes test
Standardised against C₂H₈N₂.Fe.2H₂SO₄ Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR