Natriumsulfat, wasserfrei ≥99,0%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Fluka™

Lieferant: Honeywell Chemicals

Synonyme: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt

31481-50KG 31481.9050 31481-1KG 31481-2.5KG 31481-6X1KG 31481-6X2.5KG 31481-500G
HWOL31481-50KGEA 0 EUR
HWOL31481-50KG HONE31481.9050 31481-1KG 31481-2.5KG 31481-6X1KG 31481-6X2.5KG 31481-500G
Natriumsulfat, wasserfrei ≥99,0%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Fluka™
Natriumsulfat
Formel: Na₂SO₄
Molecular Weight: 142.04 g/mol
Siedepunkt: 1429
Schmelzpunkt: 884 °C
Dichte: 2.68 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003504
CAS-Nummer: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay Min. 99.0 %
Assay (calculated to the dried substance) 99 - 101.0 %
Loss on ignition (800°C) Max. 0.5 %
Loss on drying (130°C) Max. 0.5 %
Water insoluble matter Max. 0.01 %
pH (5 %, 25 °C) 5.2 - 8.0
Free acid (as H₂SO₄) Max. 0.05 %
Free alkali (as NaOH) Max. 0.03 %
Arsenic (As) Max. 0.0001 %
Calcium (Ca) Max. 20 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 0.01 %
Magnesium (Mg) Max. 25 ppm
Zinc (Zn) Max. 0.02 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 10 ppm
Phosphate (PO₄) Max. 0.001 %
Total N Max. 5 ppm
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR