Kaliumhydroxid 45% in wässriger Lösung, BAKER ANALYZED®, elektronische Qualität für Mikroelektronik, J.T.Baker®

Lieferant: Avantor

Synonyme: Kalihydrat, Ätzkali

3149-09EA 0 EUR
3149-09
Kaliumhydroxid 45% in wässriger Lösung, BAKER ANALYZED®, elektronische Qualität für Mikroelektronik, J.T.Baker®
Kaliumhydroxid
Formel: KOH
Molecular Weight: 56,11 g/mol
MDL: MFCD00003553
CAS-Nummer: 1310-58-3
UN: 1814
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

For Microelectronic Use
Assay (KOH) 45.0 - 46.5 %
Color (APHA) ≤10
Appearance Passes Test
Density (g/mL) at 25°C 1.45 - 1.47
Identification Passes Test
Insoluble Matter ≤30 ppm
Potassium Carbonate (K₂CO₃) ≤1000 ppm
Ammonium Hydroxide Precipitate ≤30 ppm
Chloride (Cl) ≤30 ppm
Nitrogen Compounds (as N) ≤2 ppm
Phosphate (PO₄) ≤1 ppm
ACS - Sulfate (SO₄) ≤5 ppm
Trace Impurities - Aluminum (Al) ≤5.000 ppm
Trace Impurities - Chromium (Cr) ≤5.0 ppm
Trace Impurities - Copper (Cu) ≤0.050 ppm
Heavy Metals (as Ag) ≤3 ppm
Trace Impurities - Iron (Fe) ≤1 ppm
Trace Impurities - Lead (Pb) ≤2.000 ppm
Trace Impurities - Mercury (Hg) ≤0.10 ppm
Trace Impurities - Nickel (Ni) ≤1.000 ppm
Trace Impurities - Sodium (Na) ≤200.000 ppm
Trace Impurities - Zinc (Zn) ≤5.000 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR