Ethoxyquin, Supelco®

Lieferant: Merck

Synonyme: 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline, 6-Ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline, Ethoxyquin

M-Clarity™ Qualitätsstufe = MQ100

31519-250MG
31519-250MGEA 64.2 EUR
31519-250MG
Ethoxyquin, Supelco®
Ethoxyquin
Formel: C₁₄H₁₉NO
MDL: MFCD00023883
CAS-Nummer: 91-53-2
EINECS: 202-075-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=435fb7c7d098a71d803155c974bf8aa075913b9f&vid=91275dc969f36df987c0326d1df1dc0a0b39a388&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR