Ampicillin Trihydrat, Supelco®

Lieferant: Merck
Achtung

Synonyme: (2S,5R,6R)-6-((R)-2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate

M-Clarity™ Qualitätsstufe = MQ100

31591-250MG
31591-250MGEA 57.4 EUR
31591-250MG
Ampicillin Trihydrat, Supelco®
Ampicillin Trihydrat
Formel: C₁₆H₁₉N₃O₄S·3H₂O
Molecular Weight: 403,45 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00072036
CAS-Nummer: 7177-48-2
EINECS: 200-709-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ea0ee199a8ee4b9c086e63d8c1ab2a3b379d085&vid=3ab8cb99eb078748163f30a7ee4fd1627dc71541&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR