Bithionol, Supelco®

Lieferant: Merck

Synonyme: Bithionol, Bis(3,5-dichloro-2-hydroxyphenyl) sulphide, 2,2'-Thiobis(4,6-dichlorophenol), 6,6'-Thiobis(2,4-dichlorophenol)

M-Clarity™ Qualitätsstufe = MQ100

31622-250MG
31622-250MGEA 54 EUR
31622-250MG
Bithionol, Supelco®
Bithionol
Formel: C₁₂H₆Cl₄O₂S
MDL: MFCD00055727
CAS-Nummer: 97-18-7
EINECS: 202-565-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b52debab2e5258fe04f9109bce8eeb8aedfd7a56&vid=d97a8fef33f7f1a367149417b4c3399507d40adc&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR