2-Aminoazotoluol, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

31629-250MG
31629-250MGEA 427 EUR
31629-250MG
2-Aminoazotoluol, Supelco®
2-Aminoazotoluol
Formel: C₁₄H₁₅N₃
MDL: MFCD00007733
CAS-Nummer: 97-56-3
EINECS: 202-591-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5af980c51aeadee2e01be14f105718a885b406d&vid=b5537e2444d767faddd6ee4fc5ae714774cfb9bf&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR