Perdrogen® solution_SC ≥30% w/w Stabilised, Puriss. p.a. Reag. Ph. Eur., ISO zA, Fluka™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Hydrogenperoxid
31642-1L
31642-500ML
31642-5L
31642-6X1L
31642-1LEA
86.24
EUR
31642-1L
31642-500ML
31642-5L
31642-6X1L
Perdrogen® solution_SC ≥30% w/w Stabilised, Puriss. p.a. Reag. Ph. Eur., ISO zA, Fluka™
Wasserstoffperoxid
Honeywell ACS solvents for general laboratory use are:
- A value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- Suitable for less demanding applications such as routine isocratic HPLC
- Variations and decrease issues traceable to solvent contamination
- All solvents are tested meet ACS general use specifications
Formel:
H₂O₂ Molecular Weight: 34.01 g/mol Siedepunkt: 150.2 Zers. (beim Erhitzen) Schmelzpunkt: –26 °C Dichte: 1.11 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011333 CAS-Nummer: 7722-84-1 EINECS: 231-765-0 UN: 2014 ADR: 5.1 II Merck Index: 14,04798 |
Spezifikation Testergebnisse
Assay | Min. 30 % |
Non-volatile matter | Max. 0.005 % |
Residue on ignition | Max. 0.002 % |
Free acid (as H₂SO₄) | Max. 0.005 % |
Acidity | complying |
Aluminium (Al) | Max. 0.00005 % |
Arsenic (As) | Max. 0.000001 % |
Barium (Ba) | Max. 0.000005 % |
Beryllium (Be) | Max. 0.000001 % |
Bismuth (Bi) | Max. 0.00001 % |
Calcium (Ca) | Max. 0.00002 % |
Cadmium (Cd) | Max. 0.000001 % |
Cobalt (Co) | Max. 0.000001 % |
Chromium (Cr) | Max. 0.000002 % |
Copper (Cu) | Max. 0.000001 % |
Iron (Fe) | Max. 0.000005 % |
Germanium (Ge) | Max. 0.000005 % |
Potassium (K) | Max. 0.00001 % |
Lithium (Li) | Max. 0.000001 % |
Magnesium (Mg) | Max. 0.000005 % |
Manganese (Mn) | Max. 0.000001 % |
Molybdenum (Mo) | Max. 0.000002 % |
Sodium (Na) | Max. 0.00001 % |
Nickel (Ni) | Max. 0.000002 % |
Lead (Pb) | Max. 0.000001 % |
Strontium (Sr) | Max. 0.000001 % |
Titanium (Ti) | Max. 0.00001 % |
Thallium (Tl) | Max. 0.000005 % |
Vanadium (V) | Max. 0.000001 % |
Zinc (Zn) | Max. 0.000005 % |
Zirconium (Zr) | Max. 0.00001 % |
Chloride (Cl) | Max. 0.00005 % |
Phosphate (PO₄) | Max. 0.0001 % |
Sulfate (SO₄) | Max. 0.0001 % |
Total N | Max. 0.0002 % |
Organic stabilisators | Max. 0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...