Perdrogen® solution_SC ≥30% w/w Stabilised, Puriss. p.a. Reag. Ph. Eur., ISO zA, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Hydrogenperoxid

31642-1L 31642-500ML 31642-5L 31642-6X1L
31642-1LEA 86.24 EUR
31642-1L 31642-500ML 31642-5L 31642-6X1L
Perdrogen® solution_SC ≥30% w/w Stabilised, Puriss. p.a. Reag. Ph. Eur., ISO zA, Fluka™
Wasserstoffperoxid

Honeywell ACS solvents for general laboratory use are:


  • A value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested meet ACS general use specifications

Formel: H₂O₂
Molecular Weight: 34.01 g/mol
Siedepunkt: 150.2 Zers. (beim Erhitzen)
Schmelzpunkt: –26 °C
Dichte: 1.11 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011333
CAS-Nummer: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 II
Merck Index: 14,04798

Bestellen

Spezifikation Testergebnisse

Assay Min. 30 %
Non-volatile matter Max. 0.005 %
Residue on ignition Max. 0.002 %
Free acid (as H₂SO₄) Max. 0.005 %
Acidity complying
Aluminium (Al) Max. 0.00005 %
Arsenic (As) Max. 0.000001 %
Barium (Ba) Max. 0.000005 %
Beryllium (Be) Max. 0.000001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00002 %
Cadmium (Cd) Max. 0.000001 %
Cobalt (Co) Max. 0.000001 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000001 %
Iron (Fe) Max. 0.000005 %
Germanium (Ge) Max. 0.000005 %
Potassium (K) Max. 0.00001 %
Lithium (Li) Max. 0.000001 %
Magnesium (Mg) Max. 0.000005 %
Manganese (Mn) Max. 0.000001 %
Molybdenum (Mo) Max. 0.000002 %
Sodium (Na) Max. 0.00001 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.000001 %
Strontium (Sr) Max. 0.000001 %
Titanium (Ti) Max. 0.00001 %
Thallium (Tl) Max. 0.000005 %
Vanadium (V) Max. 0.000001 %
Zinc (Zn) Max. 0.000005 %
Zirconium (Zr) Max. 0.00001 %
Chloride (Cl) Max. 0.00005 %
Phosphate (PO₄) Max. 0.0001 %
Sulfate (SO₄) Max. 0.0001 %
Total N Max. 0.0002 %
Organic stabilisators Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR