Phenolphthalein 1% in Ethanol 70% pH-Indikator

Lieferant: VWR Chemicals
Gefahr

Synonyme: 3,3-Bis(4-hydroxyphenyl)phthalid, 3,3-Bis(4-hydroxyphenyl)-1(3H)-isobenzofuranon

31723.230EA 47.8 EUR
31723.230 31723.290
Phenolphthalein 1% in Ethanol 70% pH-Indikator
Phenolphthalein

  • Transition range pH 8.2-9.8

Formel: C₂₀H₁₄O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00005913
CAS-Nummer: 77-09-8
EINECS: 201-004-7
UN: 1170
ADR: 3,III

Bestellen

Spezifikation Testergebnisse

Application test passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR