Neutral mounting medium, DPX, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
317616-100ML
317616-100MLEA
51.5
EUR
317616-100ML
Neutral mounting medium, DPX, Sigma-Aldrich®
Färbemittel
Mikroskopie- und Färbereagenzien
D.P.X mounting medium is a mixture of distyrene (polystyrene), a plasticizer (tricresyl phosphate), and xylene. It facilitates the protection of stains and can dry quickly.
Bitte beachten Storage temperature: Room temperature