Trifluoressigsäure ≥99,5%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.5+%
CAS No.: 76-05-1
Molecular Formula: C2HF3O2
Formula Weight: 114.02
Physical Form: Liquid
Density: 1.480
Refractive Index: 1.2840
Sensitivity: Hygroscopic
Appearance: Colourless
Odour: Pungent
Melting Point: -15°
Boiling Point: 72-73°
Beilstein Reference: 742035
Merck Reference: 14,9681
UN No.: 2699
MDL No.: MFCD00004169
Formel:
C₂HF₃O₂ Siedepunkt: 72…73 °C Schmelzpunkt: –15 °C Dichte: 1,480 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004169 CAS-Nummer: 76-05-1 EINECS: 200-929-3 UN: 2699 ADR: 8,I Merck Index: 14,9681 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...