4-(Dimethylamino)azobenzene-4'-isothiocyanate 97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: DABITC, 4-(N,N-Dimethylamino)azobenzene-4'-isothiocyanate, 4-(4-Isothiocyanatophenylazo)-N,N-dimethylaniline, 4-((4-Isothiocyanatophenyl)diazenyl)-N,N-dimethylaniline

317802-1G 317802-250MG
317802-1GEA 555 EUR
317802-1G 317802-250MG
4-(Dimethylamino)azobenzene-4'-isothiocyanate 97%, Sigma-Aldrich®
4-(Dimethylamino)azobenzene-4'-isothiocyanate

4-(4-Isothiocyanatophenylazo)-N,N-dimethylaniline,97% (4-(Dimethylamino)azobenzene-4'-isothiocyanate; DABITC)

Formel: C₁₅H₁₄N₄S
Schmelzpunkt: 167…171 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00004812
CAS-Nummer: 7612-98-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a55e588bb27387c69b0b956a4bf8f621d7361b1e&vid=2faab9d47d89122363c0560ad6121b9b0793c953&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Conforms to Requirements
Orange to Red-Brown
Appearance (Form) Conforms to Requirements
Powder or Crystals
Infrared spectrum Conforms to Structure
Orange to Red-Brown
Carbon 61.6 - 66.0 %
Nitrogen 19.1 - 20.5 %
Purity (HPLC) ≥96.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR