N,N,N'-Trimethylethylendiamin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1-(Dimethylamino)-2-(methylamino)ethan, Dimethyl(2-(methylamino)ethyl)amin
31785.03
31785.06
31785.14
31785.03EA
30.1
EUR
31785.03
31785.06
31785.14
N,N,N'-Trimethylethylendiamin ≥98%
N,N,N'-Trimethylethylendiamin
Purity: 98%
CAS No.: 142-25-6
Molecular Formula: C5H14N2
Formula Weight: 102.18
Flash Point: 9°(49°F)
Physical Form: Liquid
Density: 0.804
Refractive Index: 1.4190
Odour: Amine-like
Boiling Point: 116-118°
UN No.: 2733
MDL No.: MFCD00014874
Formel:
C₅H₁₄N₂ Molecular Weight: 102.18 g/mol Siedepunkt: 116…118 °C Dichte: 0.804 Flammpunkt: 9 °C (49 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014874 CAS-Nummer: 142-25-6 EINECS: 205-529-2 UN: 2733 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...