3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] 98%
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
318426-1G
318426-250MG
318426-1GEA
459
EUR
318426-1G
318426-250MG
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] 98%
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide]
3,3′-dihexyloxacarbocyanine iodide (DiOC6) is a supravital lipophilic fluorochrome. This cationic dye is employed to stain mitochondria and endoplasmic reticulum in animal and plant cells. In addition, it acts as a very rapid, consistent and highly selective fluorochrome for endophytic ascomycetes in plant roots and liverworts. DiOC6 also facilitates the detection of different mitochondrial changes happening during early apoptosis.
- 3,3′-Dihexyloxacarbocyanine iodide has been used
- to selectively stain endophytic ascomycetes in plant roots and liverworts
- to visualize thrombus formation and platelet aggregate
- to visualize forchlorfenuron (FCF)-induced changes in mitochondrial morphology
Formel:
C₂₉H₃₇IN₂O₂ Schmelzpunkt: 219,..221 °C (Lit.) Lagertemperatur: Room Temperature |
MDL:
MFCD00011954 CAS-Nummer: 53213-82-4 |
|
Spezifikation Testergebnisse
Appearance (Colour) | Red |
Appearance (Form) | Powder or Chunks |
Infrared spectrum | Conforms to Structure |
Wavelength | 483 - 488 nm |
Extinction Coefficient, c = 0.01 g/L; Methanol | ≥120000 |
Carbon | 59.3 - 62.4% |
Nitrogen | 4.6 - 5.1% |
Purity (HPLC) | ≥97.5% |
Solubility (Turbidity) | Clear to Slightly Hazy |
Solubility (Colour), c = 25 mg/ml, CHCl3 | Orange |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...