Bromine (bromide-bromate) solution 0.05 M Br2 (0.1N) volumetrische Lösung, Fluka™
Lieferant: Honeywell Chemicals
|
Gefahr
|
318698-2L
318698-500ML
318698-2LEA
144.12
EUR
318698-2L
318698-500ML
Bromine (bromide-bromate) solution 0.05 M Br2 (0.1N) volumetrische Lösung, Fluka™
Bromine (bromide-bromate) solution
Used for volumetric detection of bromine.
Standards: Traceable to NIST, SRM.
Spezifikation Testergebnisse
Assay | 0.0950 - 0.1050 N |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...