Bromkresolgrün Natriumsalz 0,04% in wässriger Lösung, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
3,3',5,5'-Tetrabrom-m-kresolsulfonphthalein Natriumsalz, Bromkresolgrün wasserlöslich
M-Clarity™ Qualitätsstufe = MQ100
Bromocresol green is a sulfonephthalein dye. It is mainly used for measuring serum albumin. In acidic conditions, bromocresol green interacts with albumin, thereby resulting in an increase in absorbance at 630 nm. The change in absorbance is linked with the concentration of albumin. Bromocresol Green is also a pH indicator, changing from yellow (pH 3.8) to blue color (5.4). It is widely used as a colloidal pigment.
Formel:
C₂₁H₁₃Br₄NaO₅S Molecular Weight: 720 g/mol Schmelzpunkt: 225 Dichte: 0.981 g/ml (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00148898 CAS-Nummer: 62625-32-5 EINECS: 263-657-4 |
|
Spezifikation Testergebnisse
Appearance (Colour) |
Conforms to Requirements Dark Green to Dark Blue |
Appearance (Form) | Liquid |
Wavelength | ≥10000 |
Extinction Coefficient | ≥8000 |
Visual Interval pH 3.8 - Yellow |
Conforms to Requirements |
Visual Interval pH 4.5 - Green |
Conforms to Requirements |
Visual Interval pH 5.4 - Blue |
Conforms to Requirements |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...