Bromphenolblau 0,04% (w/w) in wässriger Lösung, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
3,3',5,5'-Tetrabromphenolsulfonphthalein, 4,4'-(3H-2,1-Benzoxathiol-3-yliden)bis[2,6-dibromphenol]-S,S-dioxid
M-Clarity™ Qualitätsstufe = MQ200
318744-500ML
318744-500MLEA
69.5
EUR
318744-500ML
Bromphenolblau 0,04% (w/w) in wässriger Lösung, Sigma-Aldrich®
Bromphenolblau
Bromophenol Blue is a tracking dye for nucleic acid or protein electrophoresis in agarose or polyacrylamide gels.
Formel:
C₁₉H₁₀Br₄O₅S Molecular Weight: 669,97 g/mol Dichte: 0,985 g/ml (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005875 CAS-Nummer: 115-39-9 EINECS: 204-086-2 |
Spezifikation Testergebnisse
Appearance (Colour) | Purple |
Appearance (Form) | Liquid |
Wave Length In Water |
307 - 313 nm |
Extinction Coefficient | ≥8000 |
Wave Length | 589 - 595 nm |
Visual Interval pH 3.0 - Yellow with Slight Green Tint |
≥75000 |
Visual Interval pH 3.4 - Green |
Pass |
Visual Interval pH 4.6 - Blue |
Pass |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...