2-Methylpyrrolidin ≥97%

Lieferant: Thermo Scientific
Gefahr

31902.03
31902.03EA 80.9 EUR
31902.03
2-Methylpyrrolidin ≥97%
2-Methylpyrrolidin

Purity: 97% CAS No.: 765-38-8 Molecular Formula: C5H11N Formula Weight: 85.15 Flash Point: 2°(37°F) Physical Form: Liquid Density: 0.834 Refractive Index: 1.4370 Odour: Amine-like UN No.: 2733 MDL No.: MFCD00014491

Formel: C₅H₁₁N
Molecular Weight: 85,15 g/mol
Dichte: 0,834
Flammpunkt: 2 °C (37 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014491
CAS-Nummer: 765-38-8
EINECS: 212-144-3
UN: 2733
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR