Weißes Mineralöl (Erdöl)

Lieferant: Thermo Scientific

Synonyme: Matrix oil, Mineral oil, Paraffin oil

31911.D9
31911.D9EA 317 EUR
31911.D9
Weißes Mineralöl (Erdöl)
Mineral oil, white (petroleum)

CAS No.: 8020-83-5 Physical Form: Liquid Appearance: Colourless Odour: Mineral-oil-like MDL No.: MFCD00131611

Flammpunkt: >100 °C (212 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00131611
CAS-Nummer: 8042-47-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR