Kaliumnitrat 99,0-100,5%, körnig USP, FCC, J.T.Baker®
Lieferant: Avantor
|
Achtung
|
Synonyme:
Kalisalpeter
3192-88EA
0
EUR
3192-88
Kaliumnitrat 99,0-100,5%, körnig USP, FCC, J.T.Baker®
Kaliumnitrat
Formel:
KNO₃ Dichte: 2,10 g/cm³ |
MDL:
MFCD00011409 CAS-Nummer: 7757-79-1 UN: 1486 ADR: 5.1,III |
Spezifikation Testergebnisse
Food GMP Manufactured Product | |
GMP Manufactured Product | |
Meets F.C.C. Requirements | |
Meets U.S.P Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Food Chemical | |
Bulk Pharmaceutical Chemical | 99.0 - 100.5 % |
USP - Assay (KNO₃) | 99.0 - 100.5 % |
USP - Arsenic (As) | ≤3 ppm |
USP - Chloride (Cl) | ≤0.03 % |
USP - Identification | Passes Test |
USP - Iron (Fe) | ≤0.001 % |
USP - Lead (Pb) (USP) | ≤0.001 % |
USP - Nitrite (NO₂) | ≤5 ppm |
USP - Sodium (Na) | ≤0.1 % |
USP - Sulfate (SO₄) | ≤0.1 % |
FCC - Assay (KNO₃) (dried basis) | 99.0 - 100.5 % |
FCC - Chlorate | Passes Test |
FCC - Identification | Passes Test |
FCC - Lead (Pb) (FCC) | ≤4 mg/kg |
FCC - Loss on Drying at 105°C | ≤1 % |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...