Kaliumnitrat 99,0-100,5%, körnig USP, Multikompendial, J.T.Baker®
Lieferant: Avantor
|
Achtung
|
Synonyme:
Kalisalpeter
3193-85EA
991.38
EUR
3193-85
Kaliumnitrat 99,0-100,5%, körnig USP, Multikompendial, J.T.Baker®
Kaliumnitrat
Formel:
KNO₃ Schmelzpunkt: 334 °C Dichte: 2,109 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011409 CAS-Nummer: 7757-79-1 EINECS: 231-818-8 UN: 1486 ADR: 5.1,III REACH: 01-2119488224-35 Merck Index: 13,07733 |
Spezifikation Testergebnisse
Food GMP Manufactured Product | |
Meets U.S.P Requirements | |
Meets F.C.C. Requirements | |
Meets B.P. Chemical Specifications | |
Meets E.P. Chemical Specifications | |
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
Bulk Food Chemical | 99.0 - 100.5 % |
USP - Assay (KNO₃) | 99.0 - 100.5 % |
USP - Arsenic (As) | ≤3 ppm |
USP - Chloride (Cl) | ≤0.03 % |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Iron (Fe) | ≤0.001 % |
USP - Lead (Pb) | ≤4 mg/kg |
USP - Nitrite (NO₂) | ≤5 ppm |
USP - Sodium (Na) | ≤0.1 % |
USP - Sulfate (SO₄) | ≤0.1 % |
JSFA - Arsenic (As) | ≤2 ppm |
JSFA - Chloride (Cl) | ≤0.021 % |
JSFA - Clarity and Color of Solution | Passes Test |
JSFA - Loss on Drying at 105°C (4 hours) | ≤1 % |
EP/BP - Assay (dried basis) | 99.0 - 101.0 % |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Acidity or Alkalinity | Passes Test |
EP/BP - Reducing Substances | Passes Test |
EP/BP - Chloride (Cl) | ≤20 ppm |
EP/BP - Sulfate (SO₄) | ≤150 ppm |
EP/BP - Ammonium (NH₄) | ≤100 ppm |
EP/BP - Calcium (Ca) | ≤100 ppm |
EP/BP - Iron (Fe) | ≤20 ppm |
EP/BP - Sodium (Na) | ≤0.10 % |
EP/BP - Loss on Drying at 105°C | ≤0.5 % |
FCC - Assay (KNO₃) (dried basis) | 99.0 - 100.5 % |
FCC - Chlorate | Passes Test |
FCC - Identification A | Passes Test |
FCC - Identification B | Passes Test |
FCC - Loss on Drying at 105°C (4 hours) | ≤1 % |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. |
|
STORAGE: Preserve in tight containers. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...