n-Octadecan ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99%
CAS No.: 593-45-3
Molecular Formula: CH3(CH2)16CH3
Formula Weight: 254.50
Flash Point: 166°(330°F)
Physical Form: Crystalline mass/melt
Density: 0.782
Refractive Index: 1.4390
Odour: Odourless
Melting Point: 28°
Boiling Point: 317°
MDL No.: MFCD00009007
Formel:
H₃C(CH₂)₁₆CH₃ Siedepunkt: 317 °C Schmelzpunkt: 28 °C Dichte: 0,782 Flammpunkt: 166 °C (330 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009007 CAS-Nummer: 593-45-3 EINECS: 209-790-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...