Ölsäure ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: cis-9-Octadecensäure

31997.06 31997.14 31997.22
31997.06EA 71.5 EUR
31997.06 31997.14 31997.22
Ölsäure ≥99%
Ölsäure

Purity: 99% CAS No.: 112-80-1 Molecular Formula: C18H34O2 Formula Weight: 282.47 Physical Form: Liquid Density: 0.887 Refractive Index: 1.4610 Sensitivity: Air Sensitive Appearance: Colourless to pale yellow Melting Point: 13-14° Boiling Point: 286°/100mm Beilstein Reference: 1726542 Merck Reference: 14,6828 MDL No.: MFCD00064242

Formel: C₁₈H₃₄O₂
Molecular Weight: 282.47 g/mol
Siedepunkt: 286 °C (100 mmHg)
Schmelzpunkt: 13…14 °C
Dichte: 0.887
Flammpunkt: 180
Lagertemperatur: Raumtemperatur
MDL: MFCD00064242
CAS-Nummer: 112-80-1
EINECS: 204-007-1
Merck Index: 13,06898
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aecc5dce4d0318386a45d7adb7c5822631010a&vid=892a2df4ecab3679b06983b84cd27ad334ef961f&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR