Ölsäure ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
cis-9-Octadecensäure
31997.06
31997.14
31997.22
31997.06EA
71.5
EUR
31997.06
31997.14
31997.22
Ölsäure ≥99%
Ölsäure
Purity: 99%
CAS No.: 112-80-1
Molecular Formula: C18H34O2
Formula Weight: 282.47
Physical Form: Liquid
Density: 0.887
Refractive Index: 1.4610
Sensitivity: Air Sensitive
Appearance: Colourless to pale yellow
Melting Point: 13-14°
Boiling Point: 286°/100mm
Beilstein Reference: 1726542
Merck Reference: 14,6828
MDL No.: MFCD00064242
Formel:
C₁₈H₃₄O₂ Molecular Weight: 282.47 g/mol Siedepunkt: 286 °C (100 mmHg) Schmelzpunkt: 13…14 °C Dichte: 0.887 Flammpunkt: 180 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064242 CAS-Nummer: 112-80-1 EINECS: 204-007-1 Merck Index: 13,06898 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...